Skip to Content
Merck

Skip To

RAB0199

Mouse Galectin-7 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

Gas6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
14 400,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Gas6(14456)
Input:
sample type plasma
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

14 400,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 65 pg/mL
standard curve range: 65.54-16000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Gas6(14456)

General description

The Mouse Galectin-3 (chemokine, interferon γinducible T cell chemoattractant) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Galectin-3 in serum, plasma and cell culture supernatants.

Immunogen

Recombinant Mouse Galectin-7

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0337RAB0336RAB1143
description

for serum, plasma and cell culture supernatant

description

for serum, plasma, cell culture supernatants and urine

description

for serum, plasma and cell culture supernatant

description

for cell culture supernatants, plasma, and serum samples

Gene Information

mouse ... Gas6(14456)

Gene Information

human ... LGALS7(3963)

Gene Information

mouse ... Lgals3(16854)

Gene Information

rat ... Lgals3(83781)

species reactivity

mouse

species reactivity

human

species reactivity

mouse

species reactivity

mouse, rat

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type plasma
sample type serum sample(s)
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 65 pg/mL
standard curve range: 65.54-16000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 13.72-10000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.19-2000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/ml

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

technique(s)

capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters