Skip to Content
Merck

RAB0282

Equine Interleukin-1 Receptor Antagonist / IL-1F3 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

IL-1ra

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
eCl@ss:
32160701
Gene information:
equine ... IL1RN(100034236)
Input:
sample type cell culture supernatant(s)
sample type serum
sample type plasma
Species reactivity:
equine
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

equine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 12.29-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

equine ... IL1RN(100034236)

General description

The Equine IL-1ra (interleukin 1 receptor antagonist) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of equine IL-1ra in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Equine IL-1 Receptor Antagonist

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Interleukin-1 receptor antagonist (IL-1RA) inhibits the activity of IL-1α and IL-1β by competitively binding to their type I and type II receptors. IL-1RA plays a vital role in inhibition of acute pro-inflammatory response and forbidding chronic inflammation from damaging healthy tissues. Abnormal expression of IL-1RA leads to development of inflammatory or autoimmune related diseases. Allelic polymorphism of IL-1Rα leads to inflammatory bowel disease (IBD). Therapeutically, IL-1Rα may help in the treatment of sepsis, rheumatoid arthritis, asthma. Overexpression of IL-1Rα is observed in serum of psoriatic arthritis (PsA) patients.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library