Skip to Content
Merck

Skip To

RAB0544

Human IL20 / Interleukin-20 ELISA Kit

for serum, Plasma, Cell Culture Supertant and urine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
14 500,00 CZK

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
eCl@ss:
32160702
Gene information:
human ... IL20(50604)
Input:
sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

14 500,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 1_pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL20(50604)

General description

This ELISA antibody pair detects human IL-20.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0812RAB0525RAB0689
description

for serum, Plasma, Cell Culture Supertant and urine

description

-

description

for serum, plasma and cell culture supernatants

description

-

Gene Information

human ... IL20(50604)

Gene Information

mouse ... Il20(58181)

Gene Information

canine ... IL6(403985)

Gene Information

human ... IL36G(56300)

species reactivity

human

species reactivity

mouse

species reactivity

canine

species reactivity

human

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 1_pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 16 pg/mL
standard curve range: 16.38-4000 pg/mL

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.1 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 13 pg/mL
standard curve range: 12.29-3000 pg/mL

input

sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters