Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Please contact Customer Service for Availability | 22 800,00 CZK |
About This Item
22 800,00 CZK
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CD97(976)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description clone MEM-180, purified immunoglobulin, buffered aqueous solution | description - | description - |
| species reactivity human | species reactivity human | species reactivity mouse | species reactivity mouse |
| Gene Information human ... CD97(976) | Gene Information human ... CD97(976) | Gene Information mouse ... Cd27(21940) | Gene Information mouse ... Cd27(21940) |
| assay range inter-assay cv: <10% | assay range - | assay range inter-assay cv: <10% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) flow cytometry: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| detection method colorimetric | detection method - | detection method - | detection method colorimetric |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




