Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Please contact Customer Service for Availability | 22 800,00 CZK |
About This Item
sample type serum
sample type cell culture supernatant(s)
22 800,00 CZK
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type plasma
sample type serum
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 16 pg/mL
standard curve range: 16.38-4000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Spint1(20732)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| species reactivity mouse | species reactivity human | species reactivity human | species reactivity mouse |
| Gene Information mouse ... Spint1(20732) | Gene Information human ... SPINT1(6692) | Gene Information human ... SPINT2(10653) | Gene Information mouse ... Ifnz(319146) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type plasma | input sample type cell culture supernatant(s) | input sample type serum | input sample type plasma |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




