Skip to Content
Merck

Skip To

RAB0843

Mouse Tnfrsf1B / Tumor Necrosis Factor Receptor Superfamily Member 1B ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
22 800,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Tnfrsf1B(21938)
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

22 800,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Tnfrsf1B(21938)

General description

The antibody pair provided in this kit recognizes mouse Tumor Necrosis Factor Receptor Superfamily Member 1B.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0855RAB0842RAB0844
description

-

description

-

description

-

description

-

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

Gene Information

mouse ... Tnfrsf1B(21938)

Gene Information

mouse ... Tnfrsf19(29820)

Gene Information

mouse ... Tnfrsf1A(21937)

Gene Information

mouse ... Tnfrsf13B(57916)

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.24 ng/mL
standard curve range: 0.246-60 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.23-300 pg/mL

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

Pictograms

Health hazard

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

10 - Combustible liquids

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters