Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 22 800,00 CZK |
About This Item
NACRES:
NA.32
UNSPSC Code:
41116158
22 800,00 CZK
Please contact Customer Service for Availability
species reactivity
human
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TNK1(8711)
General description
The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-TNK1.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| Gene Information human ... TNK1(8711) | Gene Information human ... TYK2(7297) | Gene Information human ... ALK(238) | Gene Information human ... TYRO3(7301) |
| assay range inter-assay cv: <10% | assay range inter-assay cv: <10% | assay range inter-assay cv: <10% | assay range inter-assay cv: <10% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




