Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Please contact Customer Service for Availability | 20 800,00 CZK |
About This Item
sample type cell culture supernatant(s)
sample type plasma
20 800,00 CZK
species reactivity
bovine
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/ml
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
bovine ... IL21(378475)
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for cell culture supernatants, plasma, and serum samples | description for serum, plasma and cell culture supernatant | description for cell culture supernatants, plasma, and serum samples | description for serum, plasma, cell culture supernatant and urine |
| species reactivity bovine | species reactivity mouse | species reactivity rabbit | species reactivity human |
| Gene Information bovine ... IL21(378475) | Gene Information mouse ... Il21(60505) | Gene Information rabbit ... CXCL8(100009129) | Gene Information human ... IL3(147740) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type serum sample(s) | input sample type serum | input sample type cell culture supernatant(s) | input sample type urine |
| technique(s) ELISA: suitable | technique(s) capture ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




