Skip to Content
Merck

Skip To

RAB1015

Canine Galectin-3 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
14 500,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
canine
Storage temp.:
−20°C
Shipped in:
wet ice

14 500,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

canine

technique(s)

ELISA: suitable

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/ml

shipped in

wet ice

storage temp.

−20°C

General description

This ELISA antibody pair detects Canine Galectin-3 (LGALS3)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1143RAB0336RAB1901
description

for cell culture supernatants, plasma, and serum samples

description

for cell culture supernatants, plasma, and serum samples

description

for serum, plasma and cell culture supernatant

description

for serum, plasma and cell culture supernatants

species reactivity

canine

species reactivity

mouse, rat

species reactivity

mouse

species reactivity

rat

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/ml

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.19-2000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.15 ng/mL
standard curve range: 0.102-25 ng/mL

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

capture ELISA: suitable

technique(s)

ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters