Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Please contact Customer Service for Availability | 14 700,00 CZK |
About This Item
sample type plasma
sample type cell culture supernatant(s)
14 700,00 CZK
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.2 g/mL
standard curve range: 0.200-50 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... HPX(3263)
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants |
| species reactivity human | species reactivity human | species reactivity bovine | species reactivity human |
| Gene Information human ... HPX(3263) | Gene Information human ... GAPDH(2597) | Gene Information bovine ... CXCL10(615107) | Gene Information human ... POU5F1(5460) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type serum | input sample type cell culture supernatant(s) | input sample type plasma | input sample type cell culture supernatant(s) |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Storage Class
10 - Combustible liquids
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




