Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 22 800,00 CZK |
About This Item
sample type plasma
sample type cell culture supernatant(s)
22 800,00 CZK
species reactivity
human
technique(s)
ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... RYK(6259)
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants |
| species reactivity human | species reactivity human | species reactivity rat | species reactivity rat |
| Gene Information human ... RYK(6259) | Gene Information human ... ACE(1636) | Gene Information - | Gene Information - |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type serum | input sample type serum | input - | input - |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




