Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 22 800,00 CZK |
About This Item
sample type cell culture supernatant(s)
sample type plasma
22 800,00 CZK
species reactivity
porcine
technique(s)
ELISA: suitable
input
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
Porcine ... TNFRSF12A(100217390)
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for serum, plasma and cell culture supernatants | description for cell culture supernatants, plasma, and serum samples |
| Gene Information Porcine ... TNFRSF12A(100217390) | Gene Information - | Gene Information human ... TNFSF12(407977) | Gene Information rat ... Tnfrsf12a(302965) |
| species reactivity porcine | species reactivity rabbit | species reactivity human | species reactivity rat |
| input sample type serum | input - | input sample type cell culture supernatant(s) | input sample type plasma |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



