Skip to Content
Merck

Skip To

RAB1895

Rat Eotaxin-1 ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
22 800,00 CZK

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Gene information:
rat ... Ccl11(29397)
Input:
sample type cell culture supernatant(s)
sample type plasma
sample type serum
Species reactivity:
rat
Storage temp.:
−20°C
Shipped in:
wet ice

22 800,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.64-400 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Ccl11(29397)

General description

This ELISA antibody pair detects Rat Eotaxin-1 (CCL11)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0042RAB1797RAB0043
description

for serum, plasma and cell culture supernatants

description

for serum, plasma, cell culture supernatant and urine

description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatant

Gene Information

rat ... Ccl11(29397)

Gene Information

human ... CCL11(6356)

Gene Information

-

Gene Information

mouse ... Ccl11(20292)

species reactivity

rat

species reactivity

human

species reactivity

rhesus macaque

species reactivity

mouse

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.64-400 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 45 pg/mL
standard curve range: 49.15-12000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.02-100 pg/mL

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

input

-

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

capture ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

-

detection method

colorimetric


Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters