Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
PLA, WB
Citations:
-
biological source
rabbit
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 76.1 kDa
species reactivity
human
technique(s)
proximity ligation assay: suitable, western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... RALBP1(10928)
Immunogen
RALBP1 (NP_006779.1, 1 a.a. ~ 655 a.a) full-length human protein.
Sequence
MTECFLPPTSSPSEHRRVEHGSGLTRTPSSEEISPTKFPGLYRTGEPSPPHDILHEPPDVVSDDEKDHGKKKGKFKKKEKRTEGYAAFQEDSSGDEAESPSKMKRSKGIHVFKKPSFSKKKEKDFKIKEKPKEEKHKEEKHKEEKHKEKKSKDLTAADVVKQWKEKKKKKKPIQEPEVPQIDVPNLKPIFGIPLADAVERTMMYDGIRLPAVFRECIDYVEKYGMKCEGIYRVSGIKSKVDELKAAYDREESTNLEDYEPNTVASLLKQYLRDLPENLLTKELMPRFEEACGRTTETEKVQEFQRLLKELPECNYLLISWLIVHMDHVIAKELETKMNIQNISIVLSPTVQISNRVLYVFFTHVQELFGNVVLKQVMKPLRWSNMATMPTLPETQAGIKEEIRRQEFLLNCLHRDLQGGIKDLSKEERLWEVQRILTALKRKLREAKRQECETKIAQEIASLSKEDVSKEEMNENEEVINILLAQENEILTEQEELLAMEQFLRRQIASEKEEIERLRAEIAEIQSRQQHGRSETEEYSSESESESEDEEELQIILEDLQRQNEELEIKNNHLNQAIHEEREAIIELRVQLRLLQMQRAKAEQQAQEDEEPEWRGGAVQPPRDGVLEPKAAKEQPKAGKEPAKPSPSRDRKETSI
Sequence
MTECFLPPTSSPSEHRRVEHGSGLTRTPSSEEISPTKFPGLYRTGEPSPPHDILHEPPDVVSDDEKDHGKKKGKFKKKEKRTEGYAAFQEDSSGDEAESPSKMKRSKGIHVFKKPSFSKKKEKDFKIKEKPKEEKHKEEKHKEEKHKEKKSKDLTAADVVKQWKEKKKKKKPIQEPEVPQIDVPNLKPIFGIPLADAVERTMMYDGIRLPAVFRECIDYVEKYGMKCEGIYRVSGIKSKVDELKAAYDREESTNLEDYEPNTVASLLKQYLRDLPENLLTKELMPRFEEACGRTTETEKVQEFQRLLKELPECNYLLISWLIVHMDHVIAKELETKMNIQNISIVLSPTVQISNRVLYVFFTHVQELFGNVVLKQVMKPLRWSNMATMPTLPETQAGIKEEIRRQEFLLNCLHRDLQGGIKDLSKEERLWEVQRILTALKRKLREAKRQECETKIAQEIASLSKEDVSKEEMNENEEVINILLAQENEILTEQEELLAMEQFLRRQIASEKEEIERLRAEIAEIQSRQQHGRSETEEYSSESESESEDEEELQIILEDLQRQNEELEIKNNHLNQAIHEEREAIIELRVQLRLLQMQRAKAEQQAQEDEEPEWRGGAVQPPRDGVLEPKAAKEQPKAGKEPAKPSPSRDRKETSI
Physical form
Solution in phosphate buffered saline, pH 7.4
Still not finding the right product?
Explore all of our products under Anti-RALBP1 antibody produced in rabbit
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.