Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Please contact Customer Service for Availability | 13 600,00 CZK |
About This Item
13 600,00 CZK
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 34.9 kDa
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PRSS21(10942)
General description
Immunogen
Sequence
MGARGALLLALLLARAGLRKPESQEAAPLSGPCGRRVITSRIVGGEDAELGRWPWQGSLRLWDSHVCGVSLLSHRWALTAAHCFETYSDLSDPSGWMVQFGQLTSMPSFWSLQAYYTRYFVSNIYLSPRYLGNSPYDIALVKLSAPVTYTKHIQPICLQASTFEFENRTDCWVTGWGYIKEDEALPSPHTLQEVQVAIINNSMCNHLFLKYSFRKDIFGDMVCAGNAQGGKDACFGDSGGPLACNKNGLWYQIGVVSWGVGCGRPNRPGVYTNISHHFEWIQKLMAQSGMSQPDPSWPLLFFPLLWALPLLGPV
Physical form
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description purified immunoglobulin, buffered aqueous solution | description purified immunoglobulin, buffered aqueous solution | description affinity isolated antibody | description - |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form - |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| species reactivity human | species reactivity human | species reactivity horse, rat, rabbit, human, guinea pig, bovine, dog, mouse | species reactivity rat, human, mouse |
| Gene Information human ... PRSS21(10942) | Gene Information human ... PRSS21(10942) | Gene Information human ... PRSS23(11098) | Gene Information human ... PRSS2(5645) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



