Skip to Content
Merck

Skip To

SAB1410152

Anti-WWP1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
13 600,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

13 600,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 105.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WWP1(11059)

General description

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 4 tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. The encoded protein belongs to a family of NEDD4-like proteins, which are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. Alternative splicing of this gene generates at least 6 transcript variants; however, the full length nature of these transcripts has not been defined. (provided by RefSeq)

Immunogen

WWP1 (NP_008944.1, 1 a.a. ~ 922 a.a) full-length human protein.

Sequence
MATASPRSDTSNNHSGRLQLQVTVSSAKLKRKKNWFGTAIYTEVVVDGEITKTAKSSSSSNPKWDEQLTVNVTPQTTLEFQVWSHRTLKADALLGKATIDLKQALLIHNRKLERVKEQLKLSLENKNGIAQTGELTVVLDGLVIEQENITNCSSSPTIEIQENGDALHENGEPSARTTARLAVEGTNGIDNHVPTSTLVQNSCCSYVVNGDNTPSSPSQVAARPKNTPAPKPLASEPADDTVNGESSSFAPTDNASVTGTPVVSEENALSPNCTSTTVEDPPVQEILTSSENNECIPSTSAELESEARSILEPDTSNSRSSSAFEAAKSRQPDGCMDPVRQQSGNANTETLPSGWEQRKDPHGRTYYVDHNTRTTTWERPQPLPPGWERRVDDRRRVYYVDHNTRTTTWQRPTMESVRNFEQWQSQRNQLQGAMQQFNQRYLYSASMLAAENDPYGPLPPGWEKRVDSTDRVYFVNHNTKTTQWEDPRTQGLQNEEPLPEGWEIRYTREGVRYFVDHNTRTTTFKDPRNGKSSVTKGGPQIAYERGFRWKLAHFRYLCQSNALPSHVKINVSRQTLFEDSFQQIMALKPYDLRRRLYVIFRGEEGLDYGGLAREWFFLLSHEVLNPMYCLFEYAGKNNYCLQINPASTINPDHLSYFCFIGRFIAMALFHGKFIDTGFSLPFYKRMLSKKLTIKDLESIDTEFYNSLIWIRDNNIEECGLEMYFSVDMEILGKVTSHDLKLGGSNILVTEENKDEYIGLMTEWRFSRGVQEQTKAFLDGFNEVVPLQWLQYFDEKELEVMLCGMQEVDLADWQRNTVYRHYTRNSKQIIWFWQFVKETDNEVRMRLLQFVTGTCRLPLGGFAELMGSNGPQKFCIEKVGKDTWLPRSHTCFNRLDLPPYKSYEQLKEKLLFAIEETEGFGQE

Physical form

Solution in phosphate buffered saline, pH 7.4

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2102717WH0011059M1HPA023180
description

purified immunoglobulin, buffered aqueous solution

description

affinity isolated antibody

description

clone 1A7, purified immunoglobulin, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

Gene Information

human ... WWP1(11059)

Gene Information

human ... WWP1(11059)

Gene Information

human ... WWP1(11059)

Gene Information

human ... WWP1(11059)


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters