Skip to Content
Merck

Skip To

SAB1410176

Anti-SNF8 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Dot3, EAP30, VPS22

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
13 600,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
-

13 600,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 28.9 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable, western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNF8(11267)

General description

SNF8, VPS25 (MIM 610907), and VPS36 (MIM 610903) form ESCRT-II (endosomal sorting complex required for transport II), a complex involved in endocytosis of ubiquitinated membrane proteins. SNF8, VPS25, and VPS36 are also associated in a multiprotein complex with RNA polymerase II elongation factor (ELL; MIM 600284) (Slagsvold et al., 2005 [PubMed 15755741]; Kamura et al., 2001 [PubMed 11278625]).[supplied by OMIM

Immunogen

SNF8 (NP_009172.2, 1 a.a. ~ 258 a.a) full-length human protein.

Sequence
MHRRGVGAGAIAKKKLAEAKYKERGTVLAEDQLAQMSKQLDMFKTNLEEFASKHKQEIRKNPEFRVQFQDMCATIGVDPLASGKGFWSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYLIQSVPAELNMDHTVVLQLAEKNGYVTVSEIKASLKWETERARQVLEHLLKEGLAWLDLQAPGEAHYWLPALFTDLYSQEITAEEAREALP

Physical form

Solution in phosphate buffered saline, pH 7.4

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA059320AV40071HPA034797
description

purified immunoglobulin, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

IgG fraction of antiserum

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

mol wt

antigen 28.9 kDa

mol wt

-

mol wt

55 kDa

mol wt

-

species reactivity

human

species reactivity

human

species reactivity

pig, mouse, bovine, rat, human, horse

species reactivity

human


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters