Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160703
Form:
liquid
biological source
synthetic
form
liquid
species reactivity
mouse
concentration
200 μg/mL
shipped in
wet ice
storage temp.
−20°C
Immunogen
20 amino amino terminus of mouse ICAD.
Application
Used as a blocking peptide in immunoblotting applications.
Other Notes
This peptide blocks the action of antibody PRS2001.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.