Skip to Content
Merck

Skip To

SML0910

TC3.6

≥98% (HPLC)

Synonym(s):

3-Phenyl-2,4(1H,3H)-quinazolinedithione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
8 520,00 CZK

About This Item

Empirical Formula (Hill Notation):
C14H10N2S2
CAS Number:
Molecular Weight:
270.37
UNSPSC Code:
41106305
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

8 520,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Sc1[n]([c](c3c(n1)cccc3)=S)c2ccccc2

InChI key

DJJOAJCVIYRELX-UHFFFAOYSA-N

Biochem/physiol Actions

TC3.6 is a PDE7 selective inhibitor (IC50 = 1.04 μM). In an Experimental Autoimmune Encephalomyelitis (EAE) murine model, TC3.6 reduced levels of IL-17, blocked infiltration of immune cells into the CNS and elevated levels of the Treg marker Foxp3. TC3.6 also enhances survival and differentiation of oligodendrocyte precursor cells.
TC3.6 is a PDE7 selective inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0961Y102SML0717
description

≥98% (HPLC)

description

≥98% (HPLC)

description

powder

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

-

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

room temp

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 15 mg/mL, clear

solubility

H2O: insoluble, DMSO: 10 mg/mL

solubility

H2O: 20 mg/mL, clear

color

light orange to dark orange

color

white to beige

color

white to yellow

color

white to beige


Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters