Skip to Content
Merck

Skip To

SML2267

GeA-69

≥98% (HPLC)

Synonym(s):

N-[2-(9H-Carbazol-1-yl)phenyl]-acetamide

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
7 450,00 CZK

About This Item

Empirical Formula (Hill Notation):
C20H16N2O
CAS Number:
Molecular Weight:
300.35
UNSPSC Code:
12352200
NACRES:
NA.77
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
Sign in to see your pricing

7 450,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

off-white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(NC1=CC=CC=C1C2=CC=CC3=C2NC4=CC=CC=C43)=O

InChI key

LDPDVNYCIOHPGF-UHFFFAOYSA-N

Biochem/physiol Actions

GeA-69 is a selective allosteric inhibitor of macrodomain 2 of poly-adenosine-diphosphate-ribose polymerase 14 (PARP14), an anti-apoptotic pro-survival protein associated with inflammatory diseases and several types of cancer. GeA-69 exhibited a KD of 860 nM in ITC experiments. GeA-69 was shown to be an allosteric inhibitor of adenosine diphosphate ribose (ADPR) binding to PARP14 macrodomain 2 and to prevent recruitment of the PAPP14 macrodomain 2 to DNA damage sites.
Selective allosteric inhibitor of PARP14 macrodomain 2


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters