Skip to Content
Merck

Skip To

SML2986

NSC 15520

≥98% (HPLC)

Synonym(s):

15-Carboxy-13-isopropylatis-13-ene-17,18-dioic acid, NSC15520

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
3 270,00 CZK
25 mg
Please contact Customer Service for Availability
10 500,00 CZK

About This Item

Empirical Formula (Hill Notation):
C24H34O6
CAS Number:
Molecular Weight:
418.52
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

3 270,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[P](=O)(OCC\C(=C(/N(Cc2c(nc(nc2)C)N)C=O)\C)\SC(=O)c1ccccc1)(O)O

InChI

1S/C19H23N4O6PS/c1-13(23(12-24)11-16-10-21-14(2)22-18(16)20)17(8-9-29-30(26,27)28)31-19(25)15-6-4-3-5-7-15/h3-7,10,12H,8-9,11H2,1-2H3,(H2,20,21,22)(H2,26,27,28)/b17-13+

InChI key

BTNNPSLJPBRMLZ-GHRIWEEISA-N

Biochem/physiol Actions

Inhibitor of the RPA70 N-terminal protein interaction domain
NSC15520 is an inhibitor of the N-terminal protein interaction domain of Replication Protein A (RPA), the major single-stranded DNA binding protein in eukaryotes, which is required for DNA replication, DNA repair, DNA recombination, and DNA damage response signaling. It has been shown to block the association of RPA to p53 and RAD9. In pluripotent stem cells, NSC 15520 increased targeted nucleotide substitutions in CRISPER reactions using Cas9n and Cpf1. It has been used along with a group of other small molecules to achieve a 2.8 to 7.2-fold increase in precision gene editing with Cas9.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCR060RF
description

≥98% (HPLC)

description

-

form

powder

form

-

assay

≥98% (HPLC)

assay

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

-

solubility

DMSO: 2 mg/mL, clear

solubility

-

color

white to beige

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters