Skip to Content
Merck

Skip To

SML3003

AS3334034

≥98% (HPLC)

Synonym(s):

7-Methoxy-6-(2-methylpropane-2-sulfonyl)-N-(4-methyl-1H-pyrazol-3-yl)quinolin-4-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 930,00 CZK
25 mg
Please contact Customer Service for Availability
11 700,00 CZK

About This Item

Empirical Formula (Hill Notation):
C18H22N4O3S
CAS Number:
Molecular Weight:
374.46
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

2 930,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=CNN=C1NC2=CC=NC3=CC(OC)=C(S(=O)(C(C)(C)C)=O)C=C32

InChI

1S/C18H22N4O3S/c1-11-10-20-22-17(11)21-13-6-7-19-14-9-15(25-5)16(8-12(13)14)26(23,24)18(2,3)4/h6-10H,1-5H3,(H2,19,20,21,22)

InChI key

WCPMUQAUSFXFJE-UHFFFAOYSA-N

Biochem/physiol Actions

AS3334034 is an orally available, potent and selective RIP2 (Receptor-interacting protein kinase 2) inhibitor that exhibits renoprotective effects in uninephrectomized adriamycin-induced CKD (chronic kidney disease) rats. It appears that AS3334034 suppresses the progression of chronic renal failure via an anti-inflammatory effect.
Orally available, potent and selective RIP2 inhibitor that exhibits renoprotective effects


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters