Skip to Content
Merck

Skip To

SML3290

EN4

≥98% (HPLC)

Synonym(s):

N-​[2-​(4-​Ethoxyphenoxy)​phenyl]​-​4-​[[(1-​oxo-​2-​propen-​1-​yl)​amino]​methyl]​-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
2 200,00 CZK
25 mg
Please contact Customer Service for Availability
8 880,00 CZK

About This Item

Empirical Formula (Hill Notation):
C25H24N2O4
CAS Number:
Molecular Weight:
416.47
UNSPSC Code:
12352200
MDL number:
eCl@ss:
32160701
Assay:
≥98% (HPLC)
Form:
powder

2 200,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(CNC(C=C)=O)C=C1)NC2=C(OC3=CC=C(OCC)C=C3)C=CC=C2

InChI

1S/C25H24N2O4/c1-3-24(28)26-17-18-9-11-19(12-10-18)25(29)27-22-7-5-6-8-23(22)31-21-15-13-20(14-16-21)30-4-2/h3,5-16H,1,4,17H2,2H3,(H,26,28)(H,27,29)

InChI key

DKOWGKKELPCHEG-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective covalent c-MYC inhibitor that targets cysteine 171
EN4 is a cell penetrant, potent and selective covalent c-MYC inhibitor that targets cysteine 171 located within a predicted intrinsically disordered region of MYC. EN4 inhibits MYC transcriptional activity, downregulates multiple MYC transcriptional targets, reduces MYC and MAX thermal stability, and impairs tumorigenesis.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters