Skip to Content
Merck

Skip To

SML3376

RTICBM-189

≥98% (HPLC)

Synonym(s):

N-(4-Chlorophenyl)-N′-[2-(3-chlorophenyl)ethyl]urea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H14Cl2N2O
CAS Number:
Molecular Weight:
309.19
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCCC1=CC(Cl)=CC=C1)NC2=CC=C(Cl)C=C2

InChI

1S/C15H14Cl2N2O/c16-12-4-6-14(7-5-12)19-15(20)18-9-8-11-2-1-3-13(17)10-11/h1-7,10H,8-9H2,(H2,18,19,20)

InChI key

PPKIPROVSKBYAT-UHFFFAOYSA-N

Biochem/physiol Actions

A blood brain barrier permeable, potent and selective negative allosteric modulator (NAM) of the cannabinoid CB1 receptor.
RTICBM-189 is a blood brain barrier permeable, potent and selective negative allosteric modulator (NAM) of the cannabinoid CB1 receptor. RTICBM-189 selectively attenuates reinstatement of the cocaine-seeking behavior in rats (10 mg/kg, i.p.).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1804SML3968
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to brown

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters