Skip to Content

System maintenance is scheduled for Sunday, October 11, 8:00 AM to 12:00 PM CST (Sunday, 11th October 2026, 03:00 PM to 07:00 PM CET). Quotes, price checks, and order status inquiries may be temporarily unavailable.

Merck

Skip To

SML4012

VHL Binder 30

≥97% (HPLC)

Synonym(s):

(2S,4R)-1-((S)-2-(1-Fluorocyclopropane-1-carboxamido)-3,3-di-methylbutanoyl)-4-hydroxy-N-((S)-1-(2-methyl-4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide, VHL Binder Compound 30, von Hippel−Lindau (VHL) Inhibitor Compound 30

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Please contact Customer Service for Availability
5 300,00 CZK

About This Item

Empirical Formula (Hill Notation):
C28H37FN4O4S
CAS Number:
Molecular Weight:
544.68
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥97% (HPLC)
Form:
powder

5 300,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

C[C@H](NC([C@@H]1C[C@@H](O)CN1C([C@@H](NC(C2(F)CC2)=O)C(C)(C)C)=O)=O)C3=C(C=C(C=C3)C4=C(N=CS4)C)C

InChI

1S/C28H37FN4O4S/c1-15-11-18(22-17(3)30-14-38-22)7-8-20(15)16(2)31-24(35)21-12-19(34)13-33(21)25(36)23(27(4,5)6)32-26(37)28(29)9-10-28/h7-8,11,14,16,19,21,23,34H,9-10,12-13H2,1-6H3,(H,31,35)(H,32,37)/t16-,19+,21-,23+/m0/s1

InChI key

NBHYPBDUODSQHJ-DTPUHQDNSA-N

Biochem/physiol Actions

Binds to the VHL protein with high-affinity and efficiently disrupts VHL-HIF-α interaction.



VHL Binder 30 (VHL Binder, Compound 30) is a highly potent, efficacious von Hippel−Lindau (VHL) E3 ubiquitin ligase binder with greater cell permeability, higher binding affinity (Kd = 37 & 25 nM by fluorescence polarization and surface plasmon resonance, respectively) and superior ability to induce Hypoxia-inducible factor-1α (HIF-1α) transcriptional activity (% of HIF-1α stabilization levels at 50 µM for 2 hr = 182 & 224 in HeLa & HEK293 cells; HIF-1α-OH stabilization at 50 µM for 2 hr = 263, 208 in HeLa & HEK293 cells, respectively; VH298 as the reference ligand VH298).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP2084WH0007428M1SAB4503075
description

≥97% (HPLC)

description

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

description

clone 1G12, purified immunoglobulin, buffered aqueous solution

description

affinity isolated antibody

form

powder

form

frozen liquid

form

buffered aqueous solution

form

buffered aqueous solution

assay

≥97% (HPLC)

assay

≥70% (SDS-PAGE)

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

clear colorless

color

-

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library