Skip to Content
Merck

SML4076

VU0542270

≥98% (HPLC)

Synonym(s):

N-[[4-Methyl-2-(2-thienyl)-5-thiazolyl]methyl]-N′-2-thienylurea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H13N3OS3
CAS Number:
Molecular Weight:
335.47
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
eCl@ss:
32160703
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(CNC(NC2=CC=CS2)=O)SC(C3=CC=CS3)=N1

Biochem/physiol Actions

Potent and highly selective inhibitor of vascular Kir6.1/SUR2B (vascular KATP) channels.


VU0542270 is a potent and highly selective inhibitor of vascular Kir6.1/SUR2B (vascular KATP) channels (IC50 ~ 100 nM) that show no apparent activity toward Kir6.2/SUR1 and other members of the Kir channel family. It appears that VU0542270 binds to SUR2. It has a rather short in vivo half-life (t1/2 = 0.64 hour).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library