Skip to Content
Merck

Skip To

SML4342

MMF prodrug-1c

≥95% (HPLC)

Synonym(s):

(E)-4-Oxo-pent-2-enedioic acid dimethyl ester, 1,5-Dimethyl (2E)-4-oxopent-2-enedioate; (E)-Dimethyl 4-oxopent-2-enedioate, Monomethyl fumarate Prodrug-1c, NRF2 Activator, MMF Prodrug-1c, NFE2L2 Activator MMF, Prodrug-1c

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H8O5
CAS Number:
Molecular Weight:
172.14
MDL number:
UNSPSC Code:
12352203
Assay:
≥95% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

MMF prodrug-1c is a cell permeable activator of NRF2. A site-specific monomethyl fumarate (MMF) prodrug (1c) achieves targeted delivery of MMF through a peroxide-dependent Baeyer-Villiger oxidation mechanism, selectively activating NRF2 at sites of pathological oxidative stress. Upon oral administration, MMF prodrug-1c rapidly converts to a stable glutathione S-conjugate that circulates until encountering elevated peroxide concentrations (>1 μM H2O2 or >20 μM peroxynitrite) at disease sites, where it releases therapeutic MMF while avoiding indiscriminate NRF2 activation in healthy tissues. At an optimal dose of 350 μmol/kg/day, MMF prodrug-1c demonstrates superior efficacy to conventional MMF delivery in multiple chronic pain models, reversing peripheral neuropathy, osteoarthritis, and chemotherapy-induced neuropathic pain without inducing systemic side effects like glutathione depletion or cutaneous flushing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1404123SAB5700720
description

≥95% (HPLC)

description

clone 2G7, purified immunoglobulin, buffered aqueous solution

description

-

form

powder

form

buffered aqueous solution

form

buffered aqueous solution

assay

≥95% (HPLC)

assay

-

assay

-

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

color

white to beige

color

-

color

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library