Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 mg | Please contact Customer Service for Availability | 4 540,00 CZK |
About This Item
Empirical Formula (Hill Notation):
C23H22N4O
CAS Number:
Molecular Weight:
370.45
MDL number:
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
powder
4 540,00 CZK
Please contact Customer Service for Availability
Quality Segment
assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
2-8°C
Biochem/physiol Actions
PVP-037 is a novel imidazopyrimidine (IMP) scaffold and a TLR7/8 agonist, demonstrating broad innate immune activation in primary human leukocytes at concentrations ≥1 µM, including NF-κB activation and Th1-polarizing cytokine production, notably TNF, IFN-γ and IL-1β comparable to R848 at concentrations ≥ 11 µM; it exhibits limited activity in THP-1 cells and murine/porcine primary leukocytes. In vivo, PVP-037, administered via intramuscular injection, enhances influenza and SARS-CoV-2 antigen-specific humoral responses in mice at 100 nmol doses and increases antibody titers co-administered with recombinant hemagglutinin (rHA) protein, compared to rHA alone or with alum, notably inducing a higher IgG response when formulated with rHA + PVP-037.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
