Skip to Content
Merck

Skip To

SML4396

Desthiobiotin-iodoacetamide DbGGI

≥98% (HPLC)

Synonym(s):

DbGGI, Iodoacetamide-desthiobiotin, N-(2-((2-((2-(2-iodoacetamido)ethyl)amino)-2-oxoethyl)amino)-2-oxoethyl)-6-((4R,5S)-5-methyl-2-oxoimidazolidin-4-yl)hexanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H31IN6O5
CAS Number:
Molecular Weight:
538.38
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

DbGGI is a bifunctional, cysteine-reactive ABPP probe that links an iodoacetamide warhead to a desthiobiotin tag via a flexible 10-atom chain, providing a "click-free" alternative to alkyne-tagged probes. This design enables strong, reversible capture on streptavidin beads and efficient peptide elution under mild acidic conditions, which eliminates background from copper reagents and yields a cleaner, more accurate representation of pulldown results for mass spectrometry or western blot. The probe′s principal innovation is its application in a quantitative, concentration-dependent enrichment strategy that selectively identifies hyper-reactive primary protein targets (PPTs) in cell lysates—a class often obscured in conventional high-concentration ABPP. As demonstrated in U87MG cell lysates, this approach captured 116 targets at a low concentration (10 µM) versus 193 at a high concentration (250 µM). Note: Differentially label cell lysates (2 mg/mL) with low (10 µM) and high (250 µM) DbGGI from a DMSO stock for 1 hour at 37°C. Then, capture the proteomic targets via a 4-hour incubation with streptavidin beads at room temperature.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML4170SML4392SML3775
description

≥98% (HPLC)

description

≥98% (HPLC), powder, pan-cysteine profiling tag

description

≥95%

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥95%

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to pink

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library