Skip to Content
Merck

SML4397

Azide-PEG3-Desthiobiotin

≥98% (HPLC)

Synonym(s):

(4R,5S)-Desthiobiotin-PEG3-Azide, Desthiobiotin-PEG3-Azide, Desthiobiotin-Azide, N3-DTB, DTB-PEG-N3, (4R,5S)-N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]-5-methyl-2-oxo-4-imidazolidinehexanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H34N6O5
CAS Number:
Molecular Weight:
414.50
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
(Crystal, liquid, powder, solid, semi-solid)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(Crystal, liquid, powder, solid, semi-solid)

color

colorless to yellow

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Desthiobiotin-PEG3-Azide is a trifunctional chemical probe designed for the chemoselective labeling and affinity enrichment of alkyne-modified biomolecules. Its strategic design combines a terminal azide that forms a stable triazole adduct with alkyne-tagged molecules via bio-orthogonal click chemistry—either catalyzed by Copper(I) (CuAAC) or proceeding catalyst-free with strained alkynes (SPAAC)—a hydrophilic PEG3 spacer to enhance solubility, and a desthiobiotin affinity tag. The probe′s pivotal advantage is the reversible, high-affinity binding of desthiobiotin to streptavidin (Kd ≈ 10⁻¹¹ M), which permits gentle, competitive elution that preserves the integrity of captured proteins and their interaction partners. Consequently, it serves as a central reagent in chemical proteomics for the affinity pull-down, enrichment, and subsequent mass spectrometry-based identification of alkyne-labeled biomolecules from complex cellular lysates, proving indispensable where the recovery of viable protein complexes is critical.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library