Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C15H18O2
CAS Number:
Molecular Weight:
230.30
UNSPSC Code:
12352200
eCl@ss:
32160703
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Suppressors of cytokine signaling (SOCS) proteins negatively regulate cytokine receptor signaling. The SOCS family include eight members SOCS-1 to SOCS-7 and CIS. Dehydrocostus lactone (DHE) is a sesquiterpene lactone extracted from Saussurea lappa and Aucklandia lappa. The compound possesses antifungal and antimycobacterial activity and exhibit a cytotoxic effect against HepG2, OVCAR-3, and HeLa cell lines. DHE inhibits cell proliferation not only in early-stage breast cancer MCF-7 but also in more advanced stage MDA-MB-231 cancer cells, without significant effect on H184B5F5/M10 normal mammary epithelial cells. The inhibition of breast cancer cells proliferation occurs through cell cycle blockade and apoptosis. DHE inhibits cell cycle progression and cell death through SOCS-1 and SOCS-3. Dehydrocostus lactone forms a chemical-synthetic lethal combination with subinhibitory doses of a FolE2 inhibitor trimethoprim in pathogen Burkholderia pseudomalle without annihilating the gut′s essential bacteria.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description analytical standard | description phyproof® Reference Substance | description ≥98% (HPLC) |
| form powder | form - | form solid | form powder |
| assay ≥98% (HPLC) | assay ≥98.0% (GC) | assay ≥95.0% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| storage temp. -10 to -25°C | storage temp. −20°C | storage temp. −20°C | storage temp. room temp |
| solubility DMSO: 2 mg/mL, clear | solubility - | solubility - | solubility DMSO: ≥30 mg/mL |
| color white to beige | color - | color - | color white to off-white |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



