Skip to Content
Merck

Skip To

STS0032

Saponin

BioUltra, Used as detergent and surfactant

Synonym(s):

Glycosides, Saponins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 g
Please contact Customer Service for Availability
1 690,00 CZK
250 g
Please contact Customer Service for Availability
6 900,00 CZK

About This Item

CAS Number:
MDL number:
NACRES:
NA.21
UNSPSC Code:
12161900

1 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

density

1.015-1.020 g/mL at 20 °C (5% in H2O)(lit.)

SMILES string

C[C@]1(CCC23COC4([C@H]2C1)CC[C@@H]5[C@]6(CC[C@@H](C(C6CC[C@]5([C@@]4(C[C@H]3O)C)C)(C)C)O[C@H]7[C@@H]([C@H]([C@H](CO7)O[C@H]8[C@@H]([C@H]([C@@H](CO8)O)O)O)O[C@@H]9[C@@H]([C@H]([C@@H]([C@H](O9)CO)O)O)O)O[C@H]1[C@@H]([C@H]([C@@H]([C@H](O1)CO)O)O)O[C@@H]1[C@@H]([C@H]([C@@H]([C@H](O1)CO)O)O)O)C)C=O

InChI

1S/C58H94O27/c1-52(2)29-7-11-55(5)30(8-12-58-31-15-53(3,22-62)13-14-57(31,23-77-58)32(64)16-56(55,58)6)54(29,4)10-9-33(52)82-50-46(85-51-45(40(71)37(68)27(19-61)80-51)84-49-43(74)39(70)36(67)26(18-60)79-49)44(83-48-42(73)38(69)35(66)25(17-59)78-48)28(21-76-50)81-47-41(72)34(65)24(63)20-75-47/h22,24-51,59-61,63-74H,7-21,23H2,1-6H3/t24-,25-,26-,27-,28+,29?,30-,31+,32-,33+,34+,35-,36-,37-,38+,39+,40+,41-,42-,43-,44+,45-,46-,47+,48-,49-,50+,51+,53-,54+,55-,56+,57?,58?/m1/s1

InChI key

MAEBCGDGGATMSC-OSHGGGOQSA-N

General description

Saponins, belonging to the class of steroid or triterpenoid glycosides, are widely distributed in plants, marine animals, and certain bacteria, serving as crucial compounds in various research fields, including biochemical, cell biology, and beyond. These amphipathic glycosides exhibit soap-like foaming in aqueous solutions due to their distinctive structure, featuring hydrophilic glycoside moieties combined with a lipophilic triterpene derivative.


Beyond their structural uniqueness, saponins play essential roles in biochemical processes, acting as secondary metabolites that facilitate protein penetration through cell membranes. Studies indicate their amphipathic nature and potential as hemolytic agents, contributing to their utility in immunoassays by stimulating the Th1 immune response and cytotoxic T-lymphocyte production. In cell biology, saponins find applications as nonionic surfactants, influencing cellular processes and interactions.

Moreover, saponins serve as promising components in vaccine research, functioning as adjuvants that can activate the mammalian immune system. This property, combined with their ability to solubilize low-density lipoproteins and regulate cholesterol concentration in micelles, makes saponins versatile and valuable in multidisciplinary research areas, including biochemical and cell biology research.

Application

Glycoside used as a nonionic sufactant and adjuvant in vaccine development.
Saponin has been used:
as a wetting agent to study its effects on the floatability of six plastics to permeabilize washed neutrophils for immunofluorescence as a supplement in phosphate buffer saline (PBS) for immunocytochemistry

Glycoside used as a nonionic sufactant and adjuvant in vaccine development.

Biochem/physiol Actions

Saponins are nonionic surfactant that forms oil-in-water emulsions and acts as a protective colloid. They are used for the direct determination of metals in milk by flame atomic absorption spectrophotometry.
Saponins are nonionic surfactant that forms oil-in-water emulsions and acts as a protective colloid. They are used for the direct determination of metals in milk by flame atomic absorption spectrophotometry.

Features and Benefits

Tested for trace elemental impurities
Suitable for Biochemical and Cell Biology research
Can be used with HPLC and Protein Quantification
High purity product for research applications

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8451047036SAE0073
description

BioUltra, Used as detergent and surfactant

description

used as non-ionic surfactant

description

Molecular Biology, used as non-ionic surfactant

description

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

density

1.015-1.020 g/mL at 20 °C (5% in H2O)(lit.)

density

1.015-1.020 g/mL at 20 °C (5% in H2O)(lit.)

density

1.015-1.020 g/mL at 20 °C (5% in H2O)(lit.)

density

-


Still not finding the right product?

Explore all of our products under Saponin


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library