Skip to Content
Merck

Skip To

STS0302

CALXGLUK

Synonym(s):

5,11,17-Tris{2-[4-(ß-D-glucopyranosyl)-1H-1,2,3-triazol-1-yl]ethyl}-25-monoheptoxy-26,27,28-trimethoxycalix[4]arene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
1 010,00 CZK
Please contact Customer Service for Availability
1 350,00 CZK
Please contact Customer Service for Availability
2 390,00 CZK

About This Item

Empirical Formula (Hill Notation):
C71H95N9O22
Molecular Weight:
1426.56
NACRES:
NB.34
eCl@ss:
32035490

1 010,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Non-ionic

assay

95% (HPLC)

form

powder

manufacturer/tradename

CALIXAR®

CMC

0.025 mM

solubility

water: 0.025 mM, DMSO: soluble, methanol: soluble

storage temp.

−20°C

General description

CALXGLUK is a soft proprietary stabilization detergent. CALXGLUK is a non-ionic calixarene based detergent with three polar glucose groups and a heptyl chain.

Application

CALXGLUK is a calixarene detergent that can be used to

  • Solubilize membrane proteins as GPCR
  • Stabilize membrane proteins
  • Help deliver lipid detergent mixed micelles
  • Help deliver protein detergent micelles

Features and Benefits

  • Gentle detergent
  • Not sensitive to pH variations
  • Not sensitive to ionic strength
  • Able to be combined with biological materials
  • Can be used in a buffer or aqueous solution

Legal Information

CALIXAR is a registered trademark of CALIXAR SAS FRANCE

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
STS0300STS0303STS0304
description

-

description

-

description

-

description

-

form

powder

form

powder

form

powder

form

powder

CMC

0.025 mM

CMC

2.2 mM

CMC

0.54 mM

CMC

>10 mM

solubility

DMSO: soluble, water: 0.025 mM, methanol: soluble

solubility

DMSO: soluble, water: soluble, methanol: soluble

solubility

water: 27 mM, methanol: soluble, DMSO: soluble

solubility

water: 0.5 M, DMSO: soluble, methanol: soluble

manufacturer/tradename

CALIXAR®

manufacturer/tradename

CALIXAR®

manufacturer/tradename

CALIXAR®

manufacturer/tradename

CALIXAR®

assay

95% (HPLC)

assay

95% (HPLC)

assay

95% (HPLC)

assay

-

description

Non-ionic

description

-

description

-

description

Non-ionic


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library