Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C24H47NO11
Molecular Weight:
525.63
NACRES:
NB.34
eCl@ss:
32039090
Pricing and availability is not currently available.
description
Non-ionic
assay
98% (HPLC)
form
powder
manufacturer/tradename
CALIXAR®
CMC
0.25 mM
solubility
water: 1.1 mM, DMSO: soluble, methanol: soluble
storage temp.
−20°C
General description
DDLAC is a soft proprietary stabilization detergent. DDLAC is a non-ionic detergent with a lactobionamide head group and a dodecyl chain.
Application
DDLAC is used for
- Extraction of membrane proteins
- Solubilization of membrane proteins
- Stabilization of membrane proteins
Features and Benefits
- Mild detergent
- Not sensitive to pH variations
- Not sensitive to ionic strength
- Able to be combined with biological materials
- Can be used in a buffer or aqueous solution
Legal Information
CALIXAR is a registered trademark of CALIXAR SAS FRANCE
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.