Skip to Content
Merck

STS0307

FLAC6

Synonym(s):

(4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluorononylamine) lactobionamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H28F13NO11
Molecular Weight:
717.43
NACRES:
NB.34
eCl@ss:
32039090
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Non-ionic

assay

98% (HPLC)

form

powder

manufacturer/tradename

CALIXAR®

CMC

0.56 mM ((TS))

solubility

water: 28 mM, DMSO: soluble, methanol: soluble

storage temp.

−20°C

General description

FLAC6 is a soft proprietary detergent. FLAC6 is a non-ionic amphiphilic detergent with a lactobionamide head group and a fluorinated chain.

Application

FLAC6 is used for
  • Extraction of membrane proteins
  • Solubilize membrane proteins
  • Stabilization of membrane proteins
  • Creating lipid detergents
  • Creating mixed micelles
  • Creating protein detergent micelles

Features and Benefits

  • Gentle detergent
  • Not sensitive to pH variations
  • Not sensitive to ionic strength
  • Able to be combined with biological materials
  • Can be used in a buffer or aqueous solution

Legal Information

CALIXAR is a registered trademark of CALIXAR SAS FRANCE


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library