Skip to Content
Merck

TSTOPRT

ThermaStop-RT

increases RT-PCR specificity, inhibits non-specific polymerase activity, prevents mis-priming before RT-PCR

Synonym(s):

Multiplex RT-PCR, PCR Additive, hot start DNA polymerase, hot start PCR, hotstart

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

packaging

pkg of 2500 units, pkg of 500 units

technique(s)

RT-PCR: suitable, whole genome amplification: suitable

color

opaque

shipped in

ambient

storage temp.

2-8°C

General description

ThermaStop-RT is a proprietary additive for One-Step, or Two-Step Reverse Transcriptase-PCR assays containing gene-specific primers. ThermaStop-RT interacts with the reverse transcriptase at low temperatures to reduce priming errors. ThermaStop-RT improves detection sensitivity and specific product yield by eliminating priming errors at low temperature. ThermaStop-RT acts as a Hot-start which binds and inhibits DNA polymerase activity. It also renatures the DNA polymerase upon cooling and activates its functions at lower temperatures or post-amplification.

Application

ThermaStop-RT is suitable for gene expression quantitative PCR (qPCR) and next-generation sequencing (NGS) applications.

Features and Benefits

  • It eliminates the non-specific product formation
  • Provides increased sensitivity and specificity
  • Provides higher yield and pure cDNA synthesis for conventional RT-PCR
  • It minimizes the primer-dimer formation
  • It utilizes the same cycling conditions as conventional Taq polymerase
  • Decreases the RNA secondary structure
  • Stable at low temperatures
  • Enzymes are inhibited at below 55°C
  • Compatible with several RT-PCR enzymes
  • Provides better multiplexing, efficient primer use
  • Better quantitative accuracy and monoplex and multiplex amplification sensitivity

Packaging

One unit of ThermaStop-RT is defined as the amount required for optimal performance in a one-step RT-PCR containing 50 units of reverse transcriptase (RT) and one unit of hot-start Taq Polymerase in a volume of 20 uL and a temperature of 50 degrees C.

Preparation Note

Dissolve in Tris pH 8.3 buffer

Other Notes

RT-PCR, *Please note ThermaStop-RT should not be used in combination with ThermaStop or ThermaGo.

Legal Information

ThermaGo is a trademark of ThermaGenix Inc.
ThermaStop is a trademark of ThermaGenix Inc.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library