Skip to Content
Merck

Skip To

U2005

Ultrafiltration disk membranes, Millipore®

PLTK cellulosic (regenerated cellulose) disks, MWCO 30,000 Da, diam. 25 mm

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NB.24
UNSPSC Code:
41104900
eCl@ss:
32150313


material

PLTK cellulosic (regenerated cellulose) disks

description

glycerol humectant

manufacturer/tradename

Millipore PLTK02510

diam.

25 mm

pore size

30,000 Da MWCO

General description

Ultrafiltration (UF) membranes have a well-defined pellicle structure that retains molecules and particles larger than their nominal molecular weight cut off (MWCO), while allowing lower molecular-weight solvent and solutes to pass through. Dramatic concentration of macromolecules can be achieved with UF membranes. The same high-performance Millipore membranes used in Minitan plates, Pellicon cassettes, Helicon-UF cartridges, and Prostak UF modules, these disks are configured for use in pressurized, stirred ultrafiltration cells. UF membranes in stirred cells are recommended for method development prior to scale-up, or for final concentration of samples processed through Minitan-UF or Pellicon cassette systems.
Maximum rated pressure: 100 psi, depending on the hardware the disks are used with.
Temperature limit: up to 50 °C
pH range: 1-14 for PT series; 2-13 for PL series

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library