Skip to Content
Merck

Skip To

UPBSTRT

Ultra pure BST-RT Kit

Combined Ultra pure BST-RT polymerase, Ultra pure BST-RT Reaction Buffer and Magnesium sulfate

Synonym(s):

Polymerase, Reverse Transcriptase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
Technique(s):
RT-qPCR: suitable (Isothermal amplification), cDNA synthesis: suitable
Storage temp.:
−20°C (−15°C to −25°C)
Shipped in:
dry ice
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

form

liquid

storage condition

avoid repeated freeze/thaw cycles

technique(s)

RT-qPCR: suitable (Isothermal amplification), cDNA synthesis: suitable

impurities

(DNase free, RNasefree, DNA free, NICKase free)

color

colorless

suitability

suitable for PCR (Isothermal amplification)

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

General description

BST-RT Polymerase is an engineered Bst enzyme with robust reverse transcriptase (RT) activity, making it ideal for use in loop-mediated isothermal amplification (LAMP) or RT-LAMP assays. This feature eliminates the necessity of a secondary RT enzyme when targeting amplification from RNA samples, simplifying reaction setup. BST-RT works optimally at 65°C and can be used for real time reaction monitoring with a fluorescent dye or in end point detection assays. BST-RT polymerase is tolerant to potential reaction inhibitors such as ethanol, extraction buffers, and saliva. This enzyme is also compatible with workflows that incorporate dUTP for elimination of reaction carryover contaminants.

Application

Loop-mediated isothermal amplification (LAMP) has emerged as a key alternative to polymerase chain reaction (PCR)-based techniques for amplifying nucleic acids. While conventional PCR employs a thermal cycler for repetitive cycling temperatures when amplifying, isothermal amplification techniques occur at a single and fixed temperature, allowing the use of simple,portable, and more robust instruments for fast and exponential amplification.

Features and Benefits

  • Compatible with samples containing either DNA orRNA
  • Ability to perform RT-LAMP without additional RT enzymes
  • Lacks 5′ to 3′ exonuclease domain
  • DNase free, RNase free, DNA free, and Nickase free
  • Active from 25-65 °C (optimal at 65 °C)
  • Unit Definition: One unit is the amount of enzyme required to incorporate 10 nmol of dNTP onto ssDNA template for 30 minutes at 65°C
  • Also available in stand-alone, high concentrations and glycerol-free formulations


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
UPBETNE1SRE0113UPTRHCL1
description

Combined Ultra pure BST-RT polymerase, Ultra pure BST-RT Reaction Buffer and Magnesium sulfate

description

Ultra pure, Molecular Biology grade, 5M

description

recombinant, expressed in E. coli, Salt and Inhibitor Tolerant

description

Ultra pure, Molecular Biology grade pH buffering agent, 1M

technique(s)

cDNA synthesis: suitable, RT-qPCR: suitable (Isothermal amplification)

technique(s)

qPCR: suitable

technique(s)

LAMP: suitable

technique(s)

qPCR: suitable

Quality Level

400

Quality Level

400

Quality Level

200

Quality Level

400

storage condition

avoid repeated freeze/thaw cycles

storage condition

(Cold Storage (2-8°C))

storage condition

-

storage condition

room temperature (2-30 °C)

shipped in

dry ice

shipped in

-

shipped in

dry ice

shipped in

-

storage temp.

−20°C (−15°C to −25°C)

storage temp.

-

storage temp.

−20°C

storage temp.

-

grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

Molecular Biology




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library