Skip to Content
Merck

Skip To

VIV403003

SynVivo SynTumour 3D cancer mode assay kit, microvascular network configuration, 2 um pillars

Synonym(s):

Synvivo 403003

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.84
UNSPSC Code:
60104506
Storage temp.:
room temp


packaging

case of 1 ea

manufacturer/tradename

SynVivo 403003

application(s)

3D cell culture

storage temp.

room temp

General description

10 - SMN2 C001 SynTumor 3 μm gap, 2 μm height pillar barrier chips, 100 ft Tygon Tubing, 25 pack - Slide Clamps, 50 pack - 1 mL Syringe with Luer-Lok Tip, 50 pack - 24 Gauge Needle x 0.5 in long

SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment. The system enables (a) circulation in the microvasculature, (b) transport across the vessel walls, and (c) delivery to the tumors. Starting with scans of vascular networks incorporated with interstitial and tissue/tumor spaces, the SynTumor 3D tissue model creates an in-vitro tumor microenvironment akin to a viable histological slice.

The SynTumor 3D Cancer model has the following benefits:

  • Side by side architecture enables quantitative real time visualization
  • Physiological leaky vasculature with engineered porous structures
  • Morphologically realistic in vivo based architecture
  • Physiologically realistic convective and diffusive transport
Microfluidic platform with ultra-low consumable volumes

Other Notes

LInk to Publications
application text




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library