Skip to Content
Merck

Skip To

Z-2183

ZytoLight® SPEC EWSR1/FLI1 TriCheck Probe

Green/Orange/Blue, pkg of 0.05 mL

Synonym(s):

Dual color probe, fluorochrome, probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32150313
Product Z-2183 is not currently sold in your country. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ZytoVision

packaging

pkg of 0.05 mL, pkg of 5 tests

manufacturer/tradename

ZytoVision

storage condition

protect from light

color

, Green/Orange/Blue

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight®SPEC EWSR1/FLI1 TriCheck Probe is composed of:

  • ZyGreen (excitation 503 nm/emission528 nm) labeled polynucleotides (~10ng/µl), which target sequences mappingin 22q12.2** (chr22:29,779,841-30,179,900) distal to the EWSR1 breakpoint region
  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides(~4.5 ng/µl), which target sequences mapping in 22q12.1-q12.2**(chr22:29,191,431-29,673,440) proximal to the EWSR1 breakpoint region
  • ZyBlue (excitation at 418 nm andemission 467 nm) labeled polynucleotides (~37 ng/µl), which target sequences mapping in 11q24.3**(chr11:128,707,454-129,346,602) distal to the FLI1 breakpoint region
  • Formamide based hybridization buffer
The fluorescence in situ hybridization (FISH) technique allows for the detection and visualization of specific nucleic acid sequences in cell preparations. Following DAPI counterstaining of the DNA, hybridized probe fragments are observed using a fluorescence microscope with filters tailored for the fluorochromes is used to label the FISH probe fragments.
The ZytoLight® SPEC EWSR1/FLI1 TriCheck Probe (PL141) is a probe for for the qualitative detection of rearrangements involving the humanEWSR1 gene and the human FLI1 gene

Application

  • Differentiating Ewing sarcoma/PNET from classic neuroblastoma, Wilms tumor, and rhabdomyosarcoma is crucial.
  • The t(11;22)(q24;q12.2) translocation, which juxtaposes the EWSR1 gene at 22q12.2 next to the FLI1 locus, is the most frequent translocation involving the EWSR1 gene region.
  • Translocations in the 22q12.2 region are found in 90-95% of patients with Ewing sarcoma
  • Therefore, the ZytoLight® SPEC EWSR1/FLI1 TriCheck Probe is used to detect EWSR1 rearrangements by fluorescence in situ hybridization (FISH) to confirm the presence of Ewing sarcoma/PNET.

Analysis Note

  • In an interphase nucleus without a FLI1-EWSR1 rearrangement, two green/orange fusion signals and two blue signals are expected.
  • A FLI1-EWSR1 fusion is indicated by:
    • One separate orange signal co-localizing with one blue signal.
    • One separate green signal.
  • An EWSR1 translocation not involving FLI1 is indicated by:
    • The separation of one green/orange fusion signal.
    • No co-localization of the separated orange signal with one blue signal.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

75-12-7

CAS No.

75-12-7

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library