Skip to Content
Merck

Skip To

1065221

USP

Bendamustine Hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-[5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

Empirical Formula (Hill Notation):
C16H21Cl2N3O2 ·HCl
CAS Number:
Molecular Weight:
394.72
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904

10 300,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

bendamustine

manufacturer/tradename

USP

application(s)

(Pharmaceutical small molecule)

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1065243PHR23331065301
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

certified reference material, pharmaceutical secondary standard

description

United States Pharmacopeia (USP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

application(s)

(Pharmaceutical small molecule)

application(s)

pharmaceutical
pharmaceutical
pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical
pharmaceutical
pharmaceutical (small molecule)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

API family

bendamustine

API family

bendamustine

API family

bendamustine

API family

bendamustine

agency

USP

agency

USP

agency

traceable to USP 1065221

agency

USP


Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2 - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library