Skip to Content
Merck

Skip To

1096740

USP

D-Carnitine

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H15NO3
Molecular Weight:
161.20
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

-10 to -25°C

SMILES string

C[N+](C)(C)C[C@H](CC(=O)[O-])O

InChI

InChI=1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m0/s1

InChI key

PHIQHXFUZVPYII-LURJTMIESA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
738808616737870333P
description

United States Pharmacopeia (USP) Reference Standard

description

99 atom % 13C, 98 atom % D, 97% (CP)

description

98 atom % D, 98% (CP)

description

Avanti Research™ - A Croda Brand

grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

format

neat

format

-

format

-

format

-

agency

USP

agency

-

agency

-

agency

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Avanti Research™ - A Croda Brand

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

-

application(s)

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library