Skip to Content
Merck

Skip To

1162648

USP

Beta-Damascone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-(2,6,6-Trimethyl-1-cyclohexenyl)-2-buten-1-one

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904


grade

pharmaceutical primary standard

API family

damascone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C13H20O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5,7H,6,8-9H2,1-4H3/b7-5+

InChI key

BGTBFNDXYDYBEY-FNORWQNLSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

Flash Point (°F)

212.0 °F - closed cup

Flash Point (°C)

100 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters