Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 mg | Please contact Customer Service for Availability | 42 900,00 CZK |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
dronedarone
manufacturer/tradename
USP
availability
not available in (Sales restrictions may apply)
application(s)
pharmaceutical
format
neat
General description
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical |
| format neat | format neat | format neat | format neat |
| availability not available in (Sales restrictions may apply) | availability not available in (Sales restrictions may apply) | availability - | availability not available in (Sales restrictions may apply) |
| agency USP | agency USP | agency USP | agency USP |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Repr. 2
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


