Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
agency
USP
manufacturer/tradename
USP
format
neat
storage temp.
-10 to -25°C
General description
Find answers to frequently asked questions on USP products in USP FAQs.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description United States Pharmacopeia (USP) Reference Standard | description ≥93% (HPLC) | description - |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade - | grade pharmaceutical primary standard |
| format neat | format neat | format - | format neat |
| agency USP | agency USP | agency - | agency USP |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP |
| storage temp. -10 to -25°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
No data available
Flash Point (°C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



