Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Please contact Customer Service for Availability | 15 500,00 CZK |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
guarana seed extract
manufacturer/tradename
USP
application(s)
pharmaceutical
pharmaceutical
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
Application
- Guarana Seed
- Guarana Seed Powder
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical small molecule | application(s) pharmaceutical |
| format neat | format neat | format neat | format neat |
| API family guarana seed extract | API family coix seed | API family - | API family chia seed oil |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
