Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 mg | Please contact Customer Service for Availability | 18 500,00 CZK |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
hydroflumethiazide
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
O=S1(C2=CC(S(=O)(N)=O)=C(C(F)(F)F)C=C2NCN1)=O
InChI
1S/C8H8F3N3O4S2/c9-8(10,11)4-1-5-7(2-6(4)19(12,15)16)20(17,18)14-3-13-5/h1-2,13-14H,3H2,(H2,12,15,16)
InChI key
DMDGGSIALPNSEE-UHFFFAOYSA-N
Gene Information
human ... SLC12A3(6559)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Hydroflumethiazide Tablets
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description British Pharmacopoeia (BP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename BP | manufacturer/tradename USP | manufacturer/tradename EDQM |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| API family hydroflumethiazide | API family hydroflumethiazide | API family bendroflumethiazide | API family bendroflumethiazide |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under Hydroflumethiazide
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


