Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
inositol
manufacturer/tradename
USP
format
neat
storage temp.
2-8°C
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description Pharmaceutical Secondary Standard; Certified Reference Material |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical secondary standard, certified reference material |
| format neat | format neat | format neat | format neat |
| API family inositol | API family nicotinic acid | API family nicotinamide | API family nicotinamide |
| agency USP | agency USP | agency USP | agency traceable to Ph. Eur. N0600000, USP 1462006 |
| storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C | storage temp. 2-30°C |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



