Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H5Cl3
CAS Number:
Molecular Weight:
195.47
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
32150313
grade
pharmaceutical primary standard
agency
USP
API family
oxiconazole
manufacturer/tradename
USP
format
neat
storage temp.
2-8°C
General description
Find answers to frequently asked questions on USP products in USP FAQs.
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point (°F)
235.4 °F - closed cup
Flash Point (°C)
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
