Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
agency
USP
form
powder
packaging
pkg of 30 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
O=C1OC2=CC=3OC=CC3C=C2C=C1
InChI
InChI=1S/C11H6O3/c12-11-2-1-7-5-8-3-4-13-9(8)6-10(7)14-11/h1-6H
InChI key
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description ≥99% (HPLC) | description phyproof® Reference Substance | description ≥98% (HPLC), solid |
| grade pharmaceutical primary standard | grade - | grade primary reference standard | grade - |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename PhytoLab | manufacturer/tradename - |
| application(s) pharmaceutical small molecule | application(s) - | application(s) - | application(s) - |
| form powder | form powder | form powder | form solid |
| format neat | format - | format - | format - |
| packaging pkg of 30 mg | packaging - | packaging - | packaging - |
Still not finding the right product?
Explore all of our products under Psoralen
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



